N,N-Dimethyl-3-azetidinamine dihydrochloride 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: N,N-Dimethylazetidin-3-amine dihydrochloride

OR317171-1G OR317171-5G OR317171-10G
APOSOR317171-1GEA 95.7 EUR
APOSOR317171-1G APOSOR317171-5G APOSOR317171-10G
N,N-Dimethyl-3-azetidinamine dihydrochloride 97%
N,N-Dimethyl-3-azetidinamine dihydrochloride
Formel: C₅H₁₄Cl₂N₂
Molecular Weight: 173,09 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09264378
CAS-Nummer: 124668-49-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR