Methyl-2-azetidinecarboxylate hydrochloride 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: Methyl azetidine-2-carboxylate hydrochloride

OR317227-1G OR317227-5G
APOSOR317227-1GEA 115 EUR
APOSOR317227-1G APOSOR317227-5G
Methyl-2-azetidinecarboxylate hydrochloride 95%
Methyl-2-azetidinecarboxylate hydrochloride
Formel: C₅H₁₀ClNO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD01861757
CAS-Nummer: 162698-26-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR