4-Bromo-1H-pyrrolo[2,3-b]pyridin-2(3H)-one

Lieferant: Apollo Scientific
Achtung

OR317325-1G OR317325-5G
APOSOR317325-1GEA 72.5 EUR
APOSOR317325-1G APOSOR317325-5G
4-Bromo-1H-pyrrolo[2,3-b]pyridin-2(3H)-one
4-Bromo-1H-pyrrolo[2,3-b]pyridin-2(3H)-one
Formel: C₇H₅BrN₂O
Molecular Weight: 213,0314 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12406103
CAS-Nummer: 1086064-49-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR