4-(4-Chlorophenyl)-1H-pyrazole 95+%
Lieferant: Apollo Scientific
|
Achtung
|
OR318000-1G
OR318000-10G
OR318000-5G
APOSOR318000-1GEA
208
EUR
APOSOR318000-1G
APOSOR318000-10G
APOSOR318000-5G
4-(4-Chlorophenyl)-1H-pyrazole 95+%
4-(4-Chlorophenyl)-1H-pyrazole
Formel:
C₉H₇ClN₂ Schmelzpunkt: 184…185 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03647018 CAS-Nummer: 111016-47-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...