tert-Butyl-3-amino-3-oxopropyl(benzyl)carbamate 95+%
Lieferant: Apollo Scientific
OR318003-1G
OR318003-10G
OR318003-5G
APOSOR318003-1GEA
238
EUR
APOSOR318003-1G
APOSOR318003-10G
APOSOR318003-5G
tert-Butyl-3-amino-3-oxopropyl(benzyl)carbamate 95+%
tert-Butyl-3-amino-3-oxopropyl(benzyl)carbamate
Formel:
C₁₅H₂₂N₂O₃ Molecular Weight: 278,35 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD10688278 CAS-Nummer: 1040682-30-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...