4-[(Methylamino)carbonyl]benzoic acid 95+%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-(Methylcarbamoyl)benzoic acid
OR318005-1G
OR318005-10G
OR318005-5G
APOSOR318005-1GEA
382
EUR
APOSOR318005-1G
APOSOR318005-10G
APOSOR318005-5G
4-[(Methylamino)carbonyl]benzoic acid 95+%
4-[(Methylamino)carbonyl]benzoic acid
Formel:
C₉H₉NO₃ Molecular Weight: 179.18 g/mol Schmelzpunkt: 270…272 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD10688269 CAS-Nummer: 23754-45-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...