tert-Butyl-3-aminopropyl(ethyl)carbamate 95+%

Lieferant: Apollo Scientific

Synonyme: tert-Butyl 3-aminopropyl(ethyl)carbamate

OR318006-1G OR318006-5G
APOSOR318006-1GEA 249 EUR
APOSOR318006-1G APOSOR318006-5G
tert-Butyl-3-aminopropyl(ethyl)carbamate 95+%
tert-Butyl-3-aminopropyl(ethyl)carbamate
Formel: C₁₀H₂₂N₂O₂
Molecular Weight: 202,3 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11052322
CAS-Nummer: 273409-54-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR