Thiophene-2,5-dicarboxylic acid mono-tert-butylester 95+%

Lieferant: Apollo Scientific

OR318016-1G OR318016-10G OR318016-5G
APOSOR318016-1GEA 578 EUR
APOSOR318016-1G APOSOR318016-10G APOSOR318016-5G
Thiophene-2,5-dicarboxylic acid mono-tert-butylester 95+%
Thiophene-2,5-dicarboxylic acid mono-tert-butylester
Formel: C₁₀H₁₂O₄S
Molecular Weight: 228,27 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD10688273
CAS-Nummer: 503469-83-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR