2-Mercapto-4,5,6-trimethylnicotinonitrile 95+%

Lieferant: Apollo Scientific
Achtung

OR318020-1G OR318020-25G OR318020-5G
APOSOR318020-1GEA 208 EUR
APOSOR318020-1G APOSOR318020-25G APOSOR318020-5G
2-Mercapto-4,5,6-trimethylnicotinonitrile 95+%
2-Mercapto-4,5,6-trimethylnicotinonitrile
Formel: C₉H₁₀N₂S
Schmelzpunkt: 276 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00052657
CAS-Nummer: 128917-84-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR