tert-Butyl-5-hydroxy-3,4-dihydroisoquinoline-2(1H)-carboxylate 95+%

Lieferant: Apollo Scientific
Achtung

Synonyme: tert-Butyl 5-hydroxy-3,4-dihydroisoquinoline-2(1H)-carboxylate

OR318035-1G OR318035-5G
APOSOR318035-1GEA 112 EUR
APOSOR318035-1G APOSOR318035-5G
tert-Butyl-5-hydroxy-3,4-dihydroisoquinoline-2(1H)-carboxylate 95+%
tert-Butyl-5-hydroxy-3,4-dihydroisoquinoline-2(1H)-carboxylate
Formel: C₁₄H₁₉NO₃
Molecular Weight: 249,31 g/mol
Schmelzpunkt: 159…161 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD08275037
CAS-Nummer: 216064-48-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR