4-(3-Pyridin-3-yl-1H-pyrazol-4-yl)quinoline 95+%

Lieferant: Apollo Scientific
Gefahr

OR318045-1G OR318045-10G OR318045-5G
APOSOR318045-1GEA 235 EUR
APOSOR318045-1G APOSOR318045-10G APOSOR318045-5G
4-(3-Pyridin-3-yl-1H-pyrazol-4-yl)quinoline 95+%
4-(3-Pyridin-3-yl-1H-pyrazol-4-yl)quinoline
Formel: C₁₇H₁₂N₄
Molecular Weight: 272.31 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01936757
CAS-Nummer: 849924-96-5
UN: 2811
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR