2-tert-Butyl-6-methyl-1H-indole 95+%
Lieferant: Apollo Scientific
OR318047-1G
OR318047-5G
OR318047-25G
APOSOR318047-1GEA
42.2
EUR
APOSOR318047-1G
APOSOR318047-5G
APOSOR318047-25G
2-tert-Butyl-6-methyl-1H-indole 95+%
2-tert-Butyl-6-methyl-1H-indole
Formel:
C₁₃H₁₇N Lagertemperatur: Raumtemperatur |
MDL:
MFCD11052309 CAS-Nummer: 1049676-92-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...