2-tert-Butyl-4-methyl-1H-indole 95+%
Lieferant: Apollo Scientific
OR318048-1G
OR318048-5G
APOSOR318048-1GEA
893
EUR
APOSOR318048-1G
APOSOR318048-5G
2-tert-Butyl-4-methyl-1H-indole 95+%
2-tert-Butyl-4-methyl-1H-indole
Formel:
C₁₃H₁₇N Molecular Weight: 187.29 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD08275975 CAS-Nummer: 57880-12-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...