3-Amino-4-ethoxybenzoesäure 95+%

Lieferant: Apollo Scientific
Achtung

OR318056-1G OR318056-25G OR318056-5G
APOSOR318056-1GEA 235 EUR
APOSOR318056-1G APOSOR318056-25G APOSOR318056-5G
3-Amino-4-ethoxybenzoesäure 95+%
3-Amino-4-ethoxybenzoic acid
Formel: C₉H₁₁NO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD11052311
CAS-Nummer: 875256-49-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR