1-(4-Methoxybenzyl)piperidin-4-one 90%
Lieferant: Apollo Scientific
|
Achtung
|
OR318062-1G
OR318062-25G
OR318062-5G
APOSOR318062-1GEA
273
EUR
APOSOR318062-1G
APOSOR318062-25G
APOSOR318062-5G
1-(4-Methoxybenzyl)piperidin-4-one 90%
1-(4-Methoxybenzyl)piperidin-4-one
Formel:
C₁₃H₁₇NO₂ Lagertemperatur: Kühlschrank |
MDL:
MFCD09938405 CAS-Nummer: 905986-94-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...