3-Imidazol-1-yl-phenylamine

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-(1H-Imidazol-1-yl)aniline

OR318066-1G OR318066-10G OR318066-5G
APOSOR318066-1GEA 0 EUR
APOSOR318066-1G APOSOR318066-10G APOSOR318066-5G
3-Imidazol-1-yl-phenylamine
3-Imidazol-1-ylphenylamine
Formel: C₉H₉N₃
Molecular Weight: 159.19 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09258808
CAS-Nummer: 112677-67-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR