tert-Butyl-3-methyl-4-oxopiperidine-1-carboxylate 95%
Lieferant: Apollo Scientific
Synonyme:
tert-Butyl 3-methyl-4-oxopiperidine-1-carboxylate
OR318070-5G
OR318070-10G
APOSOR318070-5GEA
37.7
EUR
APOSOR318070-5G
APOSOR318070-10G
tert-Butyl-3-methyl-4-oxopiperidine-1-carboxylate 95%
tert-Butyl-3-methyl-4-oxopiperidine-1-carboxylate
Formel:
C₁₁H₁₉NO₃ Molecular Weight: 213,2735 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD08460962 CAS-Nummer: 181269-69-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...