4-[(4,6-Dimethylpyrimidin-2-yl)oxy]aniline 95+%

Lieferant: Apollo Scientific

OR318072-1G OR318072-5G
APOSOR318072-1GEA 273 EUR
APOSOR318072-1G APOSOR318072-5G
4-[(4,6-Dimethylpyrimidin-2-yl)oxy]aniline 95+%
4-[(4,6-Dimethylpyrimidin-2-yl)oxy]aniline
Formel: C₁₂H₁₃N₃O
Lagertemperatur: Raumtemperatur
MDL: MFCD01895360
CAS-Nummer: 55749-90-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR