5-Chloro-1,6-naphthyridine 95+%

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-Chloro[1,6]naphthyridine

OR318078-1G OR318078-10G OR318078-5G
APOSOR318078-1GEA 165 EUR
APOSOR318078-1G APOSOR318078-10G APOSOR318078-5G
5-Chloro-1,6-naphthyridine 95+%
5-Chloro-1,6-naphthyridine
Formel: C₈H₅ClN₂
Lagertemperatur: Raumtemperatur
MDL: MFCD01646205
CAS-Nummer: 23616-32-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR