Methyl-2-amino-4-(methoxymethyl)-1,3-thiazole-5-carboxylate 95+%

Lieferant: Apollo Scientific

OR318102-1G OR318102-10G OR318102-5G
APOSOR318102-1GEA 348 EUR
APOSOR318102-1G APOSOR318102-10G APOSOR318102-5G
Methyl-2-amino-4-(methoxymethyl)-1,3-thiazole-5-carboxylate 95+%
Methyl-2-amino-4-(methoxymethyl)-1,3-thiazole-5-carboxylate
Formel: C₇H₁₀N₂O₃S
Molecular Weight: 202,23 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12028044
CAS-Nummer: 733754-08-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR