Ethyl-2-amino-5-propylthiophene-3-carboxylate 95+%

Lieferant: Apollo Scientific

OR318103-1G OR318103-10G OR318103-5G
APOSOR318103-1GEA 288 EUR
APOSOR318103-1G APOSOR318103-10G APOSOR318103-5G
Ethyl-2-amino-5-propylthiophene-3-carboxylate 95+%
Ethyl-2-amino-5-propylthiophene-3-carboxylate
Formel: C₁₀H₁₅NO₂S
Molecular Weight: 213,3 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01922211
CAS-Nummer: 76575-31-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR