1-Acetyl-5-bromoindolin-7-amine 95+%
Lieferant: Apollo Scientific
OR318106-1G
OR318106-5G
APOSOR318106-1GEA
459
EUR
APOSOR318106-1G
APOSOR318106-5G
1-Acetyl-5-bromoindolin-7-amine 95+%
1-Acetyl-5-bromoindolin-7-amine
Formel:
C₁₀H₁₁BrN₂O Lagertemperatur: Raumtemperatur |
MDL:
MFCD09258794 CAS-Nummer: 133433-62-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...