2-Methyl-5-(methylsulphonyl)aniline 95+%

Lieferant: Apollo Scientific

OR318114-1G
APOSOR318114-1GEA 66.7 EUR
APOSOR318114-1G
2-Methyl-5-(methylsulphonyl)aniline 95+%
2-Methyl-5-(methylsulphonyl)aniline
Formel: C₈H₁₁NO₂S
Molecular Weight: 185,25 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09258802
CAS-Nummer: 1671-48-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR