2-Methyl-2-pyridin-4-ylpropanenitrile 95+%

Lieferant: Apollo Scientific
Achtung

OR318118-1G OR318118-5G OR318118-10G
APOSOR318118-1GEA 410 EUR
APOSOR318118-1G APOSOR318118-5G APOSOR318118-10G
2-Methyl-2-pyridin-4-ylpropanenitrile 95+%
2-Methyl-2-pyridin-4-ylpropanenitrile
Formel: C₉H₁₀N₂
Molecular Weight: 146.19 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07644551
CAS-Nummer: 79757-31-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR