4,4-Dimethylcyclohex-2-en-1-on-2-ylboronsäurepinakolester
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4,4-Dimethyl-2-cyclohexen-1-one-2-boronic acid pinacol ester, 4,4-Dimethyl-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)cyclohex-2-enone
OR3187-100MG
OR3187-50MG
APOSOR3187-100MGEA
55.1
EUR
APOSOR3187-100MG
APOSOR3187-50MG
4,4-Dimethylcyclohex-2-en-1-on-2-ylboronsäurepinakolester
4,4-Dimethylcyclohex-2-en-1-on-2-ylboronsäurepinakolester
Formel:
C₁₄H₂₃BO₃ Molecular Weight: 250,1400 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09027074 CAS-Nummer: 219489-09-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...