2-Brombenzaldehydoxim 96%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Bromobenzaldoxime
OR3195-25G
OR3195-5G
APOSOR3195-25GEA
487
EUR
APOSOR3195-25G
APOSOR3195-5G
2-Brombenzaldehydoxim 96%
2-Brombenzaldehydoxim
Formel:
C₇H₆BrNO Molecular Weight: 200,03 g/mol Schmelzpunkt: 102…104 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00082734 CAS-Nummer: 34158-72-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...