3-(Dimethylamino)-2-(2-thienyl)acrylonitrile 95%
Lieferant: Apollo Scientific
Synonyme:
(2Z)-3-(Dimethylamino)-2-(thiophen-2-yl)prop-2-enenitrile
OR32006-1G
OR32006-25G
OR32006-5G
APOSOR32006-1GEA
122
EUR
APOSOR32006-1G
APOSOR32006-25G
APOSOR32006-5G
3-(Dimethylamino)-2-(2-thienyl)acrylonitrile 95%
3-(Dimethylamino)-2-(2-thienyl)acrylonitrile
Formel:
C₉H₁₀N₂S Molecular Weight: 178,26 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00138239 CAS-Nummer: 165404-37-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...