5,7-Dimethyl-4-hydroxypyrido[2,3-d]pyrimidine 95%
Lieferant: Apollo Scientific
Synonyme:
5,7-DIMETHYLPYRIDO[2,3-D]PYRIMIDIN-4(3H)-ONE
OR32009-1G
OR32009-5G
APOSOR32009-1GEA
339
EUR
APOSOR32009-1G
APOSOR32009-5G
5,7-Dimethyl-4-hydroxypyrido[2,3-d]pyrimidine 95%
5,7-Dimethyl-4-hydroxypyrido[2,3-d]pyrimidine
Formel:
C₉H₉N₃O Molecular Weight: 175,19 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00143415 CAS-Nummer: 1913-72-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...