3-Chloro-N-(4-morpholinophenyl)propanamide 95%
Lieferant: Apollo Scientific
OR32039-1G
OR32039-5G
APOSOR32039-1GEA
270
EUR
APOSOR32039-1G
APOSOR32039-5G
3-Chloro-N-(4-morpholinophenyl)propanamide 95%
3-Chloro-N-(4-morpholinophenyl)propanamide
Formel:
C₁₃H₁₇ClN₂O₂ Molecular Weight: 268,75 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00172701 CAS-Nummer: 250714-83-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...