1-(2-Bromethoxy)-4-methoxybenzol 95%

Lieferant: Apollo Scientific

OR32047-1G OR32047-25G OR32047-5G
APOSOR32047-1GEA 180 EUR
APOSOR32047-1G APOSOR32047-25G APOSOR32047-5G
1-(2-Bromethoxy)-4-methoxybenzol 95%
1-(2-Bromoethoxy)-4-methoxybenzene
Formel: C₉H₁₁BrO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00017893
CAS-Nummer: 22921-76-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR