5-Chlorpicolinsäure 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-Chloro-2-pyridinecarboxylic acid, 5-Chloropyridine-2-carboxylic acid, 5-Chloro-2-picolinic acid

OR3205-25G OR3205-5G OR3205-1G
APOSOR3205-25GEA 60.9 EUR
APOSOR3205-25G APOSOR3205-5G APOSOR3205-1G
5-Chlorpicolinsäure 98%
5-Chlorpicolinsäure
Formel: C₆H₄ClNO₂
Molecular Weight: 157,56 g/mol
Schmelzpunkt: 166…171 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD04114192
CAS-Nummer: 86873-60-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR