3-(Methylsulphonyl)benzylamine hydrochloride

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-(Methylsulphonyl)benzylamine Hydrochloride

OR3206-1G OR3206-250MG
APOSOR3206-1GEA 169 EUR
APOSOR3206-1G APOSOR3206-250MG
3-(Methylsulphonyl)benzylamine hydrochloride
3-(Methylsulphonyl)benzylamine hydrochloride
Formel: C₈H₁₂ClNO₂S
Schmelzpunkt: 248…254 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09064909
CAS-Nummer: 855267-50-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR