5-Acetyl-2-chloro-6-methylnicotinonitrile 97%
Lieferant: Apollo Scientific
Synonyme:
5-Acetyl-2-chloro-6-methylpyridine-3-carbonitrile
OR32063-1G
OR32063-5G
APOSOR32063-1GEA
306
EUR
APOSOR32063-1G
APOSOR32063-5G
5-Acetyl-2-chloro-6-methylnicotinonitrile 97%
5-Acetyl-2-chloro-6-methylnicotinonitrile
Formel:
C₉H₇ClN₂O Molecular Weight: 194.62 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD01315263 CAS-Nummer: 121348-15-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...