Ethyl-4-chloro-2-phenyl-5-pyrimidinecarboxylate 95%

Lieferant: Apollo Scientific

Synonyme: Ethyl 4-chloro-2-phenylpyrimidine-5-carboxylate

OR32087-1G OR32087-5G
APOSOR32087-1GEA 244 EUR
APOSOR32087-1G APOSOR32087-5G
Ethyl-4-chloro-2-phenyl-5-pyrimidinecarboxylate 95%
Ethyl-4-chloro-2-phenyl-5-pyrimidinecarboxylate
Formel: C₁₃H₁₁ClN₂O₂
Molecular Weight: 262.7 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01935974
CAS-Nummer: 24755-82-6

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR