N'-(4-Formyl-3-methyl-1-phenyl-1H-pyrazol-5-yl)-N,N-dimethyliminoformamide 95%

Lieferant: Apollo Scientific

OR32099-1G OR32099-5G
APOSOR32099-1GEA 336 EUR
APOSOR32099-1G APOSOR32099-5G
N'-(4-Formyl-3-methyl-1-phenyl-1H-pyrazol-5-yl)-N,N-dimethyliminoformamide 95%
N'-(4-Formyl-3-methyl-1-phenyl-1H-pyrazol-5-yl)-N,N-dimethyliminoformamide
Formel: C₁₄H₁₆N₄O
Molecular Weight: 256,31 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03152811
CAS-Nummer: 58668-41-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR