3-Brom-4-methoxybenzylalkohol 98%

Lieferant: Apollo Scientific

Synonyme: (3-Bromo-4-methoxyphenyl)methanol

OR321003-25G
APOSOR321003-25GEA 224 EUR
APOSOR321003-25G
3-Brom-4-methoxybenzylalkohol 98%
3-Brom-4-methoxybenzylalkohol
Formel: C₈H₉BrO₂
Molecular Weight: 217,06 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01861398
CAS-Nummer: 38493-59-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR