(6-Bromo-1,3-benzodioxol-5-yl)methanol 95%
Lieferant: Apollo Scientific
Synonyme:
(6-Bromobenzo[d][1,3]dioxol-5-yl)methanol
OR32105-1G
OR32105-5G
APOSOR32105-1GEA
180
EUR
APOSOR32105-1G
APOSOR32105-5G
(6-Bromo-1,3-benzodioxol-5-yl)methanol 95%
(6-Bromo-1,3-benzodioxol-5-yl)methanol
Formel:
C₈H₇BrO₃ Molecular Weight: 231.05 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD02082027 CAS-Nummer: 6642-34-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...