3-Methoxy-4-(2-propynyloxy)benzenecarbaldehydeoxime 95%
Lieferant: Apollo Scientific
Synonyme:
(E)-N-{[3-Methoxy-4-(prop-2-yn-1-yloxy)phenyl]methylidene}hydroxylamine
OR32117-1G
OR32117-5G
APOSOR32117-1GEA
336
EUR
APOSOR32117-1G
APOSOR32117-5G
3-Methoxy-4-(2-propynyloxy)benzenecarbaldehydeoxime 95%
3-Methoxy-4-(2-propynyloxy)benzenecarbaldehydeoxime
Formel:
C₁₁H₁₁NO₃ Molecular Weight: 205,22 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD03001221 CAS-Nummer: 385383-46-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...