(S)-1-Boc-3-propylpiperazin 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: tert-Butyl (S)-3-propylpiperazine-1-carboxylate

OR321302-1G OR321302-100MG OR321302-250MG
APOSOR321302-1GEA 290 EUR
APOSOR321302-1G APOSOR321302-100MG APOSOR321302-250MG
(S)-1-Boc-3-propylpiperazin 95%
(S)-1-Boc-3-propylpiperazin
Formel: C₁₂H₂₄N₂O₂
Lagertemperatur: Kühlschrank
MDL: MFCD07772106
CAS-Nummer: 928025-58-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR