3-Carbamoylpyrrolidine-1-carboxylic acid tert-butyl ester 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1-(tert-Butoxycarbonyl)-3-pyrrolidinecarboxamide, 1-Boc-3-pyrrolidinecarboxamide, tert-Butyl 3-Carbamoylpyrrolidine-1-carboxylate, 3-Carbamoylpyrrolidine-1-carboxylic acid tert-Butyl ester, 1-Boc-3-Carbamoylpyrrolidine
OR321355-5G
APOSOR321355-5GEA
203
EUR
APOSOR321355-5G
3-Carbamoylpyrrolidine-1-carboxylic acid tert-butyl ester 97%
3-Carbamoylpyrrolidine-1-carboxylic acid tert-butyl ester
Formel:
C₁₀H₁₈N₂O₃ Molecular Weight: 214,27 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD04116210 CAS-Nummer: 122684-34-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...