(2R,4R)-Ethyl-4-methylpiperidine-2-carboxylate 95%

Lieferant: Apollo Scientific

Synonyme: (2R,4R)-Ethyl 4-methylpiperidine-2-carboxylate

OR321418-5G
APOSOR321418-5GEA 200 EUR
APOSOR321418-5G
(2R,4R)-Ethyl-4-methylpiperidine-2-carboxylate 95%
(2R,4R)-Ethyl-4-methylpiperidine-2-carboxylate
Formel: C₉H₁₇NO₂
Molecular Weight: 171,2368 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03093749
CAS-Nummer: 74892-82-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR