(S)-1-Boc-3-ethylpiperazin 95%

Lieferant: Apollo Scientific
Achtung

OR321450-25G OR321450-5G
APOSOR321450-25GEA 0 EUR
APOSOR321450-25G APOSOR321450-5G
(S)-1-Boc-3-ethylpiperazin 95%
(S)-1-Boc-3-ethylpiperazin
Formel: C₁₁H₂₂N₂O₂
Flammpunkt: 127 °C
Lagertemperatur: Kühlschrank
MDL: MFCD07772110
CAS-Nummer: 928025-56-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR