2,4-Dimethoxypyrimidine-5-carbaldehyde
Lieferant: Apollo Scientific
|
Achtung
|
OR3216-1G
OR3216-5G
APOSOR3216-1GEA
75.4
EUR
APOSOR3216-1G
APOSOR3216-5G
2,4-Dimethoxypyrimidine-5-carbaldehyde
2,4-Dimethoxypyrimidine-5-carbaldehyde
Formel:
C₇H₈N₂O₃ Molecular Weight: 168,15 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00132992 CAS-Nummer: 52606-02-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...