(2E)-3-(Dimethylamino)-1-(4-methoxyphenyl)prop-2-en-1-one 95%

Lieferant: Apollo Scientific

OR32176-25G OR32176-5G
APOSOR32176-25GEA 416 EUR
APOSOR32176-25G APOSOR32176-5G
(2E)-3-(Dimethylamino)-1-(4-methoxyphenyl)prop-2-en-1-one 95%
(2E)-3-(Dimethylamino)-1-(4-methoxyphenyl)prop-2-en-1-one
Formel: C₁₂H₁₅NO₂
Molecular Weight: 205,26 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00097914
CAS-Nummer: 18096-70-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR