4-Chlorobenzenecarbaldehyde-N,N-dimethylhydrazone 95%
Lieferant: Apollo Scientific
Synonyme:
(E)-2-[(4-Chlorophenyl)methylidene]-1,1-dimethylhydrazine
OR32189-1G
OR32189-25G
OR32189-5G
APOSOR32189-1GEA
180
EUR
APOSOR32189-1G
APOSOR32189-25G
APOSOR32189-5G
4-Chlorobenzenecarbaldehyde-N,N-dimethylhydrazone 95%
4-Chlorobenzenecarbaldehyde-N,N-dimethylhydrazone
Formel:
C₉H₁₁ClN₂ Molecular Weight: 182,65 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD01011933 CAS-Nummer: 22699-29-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...