3-(2-Chlorphenyl)-5-methyl-4-isoxazolcarbonsäureamid

Lieferant: Apollo Scientific

Synonyme: 3-(2-Chlorophenyl)-5-methylisoxazole-4-carbonyl chloride

OR322003-5G OR322003-25G
APOSOR322003-5GEA 0 EUR
APOSOR322003-5G APOSOR322003-25G
3-(2-Chlorphenyl)-5-methyl-4-isoxazolcarbonsäureamid
3-(2-Chlorphenyl)-5-methyl-4-isoxazolcarbonsäureamid
Formel: C₁₁H₇Cl₂NO₂
Molecular Weight: 256.09 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00020811
CAS-Nummer: 25629-50-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR