1-(4-Methylbenzyl)-1H-indole-2,3-dione 95%

Lieferant: Apollo Scientific

Synonyme: 1-[(4-Methylphenyl)methyl]-2,3-dihydro-1H-indole-2,3-dione

OR32208-1G OR32208-5G
APOSOR32208-1GEA 180 EUR
APOSOR32208-1G APOSOR32208-5G
1-(4-Methylbenzyl)-1H-indole-2,3-dione 95%
1-(4-Methylbenzyl)-1H-indole-2,3-dione
Formel: C₁₆H₁₃NO₂
Molecular Weight: 251,29 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00141570
CAS-Nummer: 79183-26-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR