1-[3-(4-Chlorophenyl)-5-isoxazolyl]-1-ethanone 97%
Lieferant: Apollo Scientific
OR32211-1G
OR32211-5G
APOSOR32211-1GEA
306
EUR
APOSOR32211-1G
APOSOR32211-5G
1-[3-(4-Chlorophenyl)-5-isoxazolyl]-1-ethanone 97%
1-[3-(4-Chlorophenyl)-5-isoxazolyl]-1-ethanone
Formel:
C₁₁H₈ClNO₂ Molecular Weight: 221,64 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00662755 CAS-Nummer: 37091-33-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...