4-(N-Boc-aminomethyl)aniline

Lieferant: Apollo Scientific

Synonyme: 4-(N-Boc-Aminomethyl)aniline

OR322111-5G OR322111-25G
APOSOR322111-5GEA 72.5 EUR
APOSOR322111-5G APOSOR322111-25G
4-(N-Boc-aminomethyl)aniline
4-(N-Boc-aminomethyl)aniline
Formel: C₁₂H₁₈N₂O₂
Molecular Weight: 222.288 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03001716
CAS-Nummer: 94838-55-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR