1-Allyl-1H-indole-2,3-dione 95%

Lieferant: Apollo Scientific

Synonyme: 1-Allyl-2,3-indolinedione, 1-Allylindole-2,3-dione, 1-Allylisatin

OR32212-1G OR32212-5G
APOSOR32212-1GEA 157 EUR
APOSOR32212-1G APOSOR32212-5G
1-Allyl-1H-indole-2,3-dione 95%
1-Allyl-1H-indole-2,3-dione
Formel: C₁₁H₉NO₂
Molecular Weight: 187,2 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00224218
CAS-Nummer: 830-74-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR